Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description: The human soluble IL-2 receptor alpha chain (sIL-2Ra) is generated by proteolytic cleavage of the cell-surface bound receptor on activated T cells. The IL-2Ra (also known as CD25) is expressed by early progenitors of the T and B lineage, as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 with low affinity. However, association with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) results in formation of the high affinity IL-2R. CD25 plays a role in lymphocyte differentiation and activation/proliferation. sIL-2Ra has been detected in the serum of patients with autoimmune disease and leukemia. Reports have demonstrated that addition of sIL-2Ra can inhibit IL-2-mediated signaling and enhance Th17 responses.
Applications Reported: Recombinant human soluble IL-2Ra is biologically active and can enhance proliferation of mouse CTLL-2 cells stimulated with 1 ng/mL IL-2.
Applications Tested: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1 ng/mL IL-2, is 0.5-1.5 µg/mL.
Source: Insect cells (amino acids Glu22-Gln240; accession #: NP_000408.1).
Bioactivity: The ED50 of this protein, as determined by increased proliferation of mouse CTLL-2 cells stimulated with 1 ng/mL IL-2, is 0.5-1.5 µg/mL.
Endotoxin: Less than 0.1 ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE and HPLC.
Molecular Weight: 24.8 kDa.
Storage and handling: For best recovery, quick spin vial prior to opening. Reconstitute in 1X PBS, pH 7.2 to a concentration of 0.1-1.0 mg/mL. Do not vortex. Use in sterile environment. Store lyophilized protein at less than or equal to -20°C. Reconstituted protein solution can be stored with carrier-protein (e.g., 0.1% BSA) at less than or equal to -20°C.
CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support