Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Entrez Gene ID: | 79621 |
Gene Name: | ribonuclease H2 subunit B |
Gene Aliases: |
AGS2, DLEU8 |
Location: |
Chr.13:50909678-50970462 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNASEH2B | NM_001142279.2 | NP_001135751.1 | ||
XM_011535229.1 | XP_011533531.1 | |||
XM_017020747.1 | XP_016876236.1 | |||
AK124228.1 | ||||
BX328027.1 | ||||
BX330672.1 | ||||
CA309004.1 |
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss | DLEU1-AS1 MIR3613 INTS6-AS1 FAM124A SPRYD7 INTS6 MIR16-1 SERPINE3 ST13P4 DLEU7-AS1 TRIM13 GUCY1B2 RNASEH2B DLEU2 MIR15A DLEU7 RNASEH2B-AS1 KCNRG DLEU1 MIR5693 LINC00371 |
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |