Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HTR2B
Transcript:
Assay Name:
Assay ID
CRISPR1026973_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
positive regulation of cytokine production
positive regulation of endothelial cell proliferation
G-protein coupled receptor internalization
heart morphogenesis
cardiac muscle hypertrophy
cGMP biosynthetic process
cellular calcium ion homeostasis
G-protein coupled receptor signaling pathway
activation of phospholipase C activity
protein kinase C-activating G-protein coupled receptor signaling pathway
serotonin receptor signaling pathway
behavior
positive regulation of cell proliferation
negative regulation of autophagy
positive regulation of phosphatidylinositol biosynthetic process
neural crest cell differentiation
phosphatidylinositol 3-kinase signaling
intestine smooth muscle contraction
phosphorylation
calcium-mediated signaling
hepatic stellate cell activation
vasoconstriction
response to drug
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAP kinase activity
positive regulation of GTPase activity
inositol phosphate metabolic process
embryonic morphogenesis
positive regulation of cytokine secretion
regulation of behavior
positive regulation of nitric-oxide synthase activity
release of sequestered calcium ion into cytosol
positive regulation of cell division
negative regulation of cell death
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
protein kinase C signaling
cellular response to amine stimulus
cellular response to temperature stimulus
cellular response to serotonin
G-protein alpha-subunit binding
G-protein coupled serotonin receptor activity
GTPase activator activity
drug binding
serotonin binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0