Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PIK3R1
Transcript:
Assay Name:
Assay ID
CRISPR1124549_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
enzyme modulator
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
cellular glucose homeostasis
negative regulation of cell-matrix adhesion
protein phosphorylation
phosphatidylinositol biosynthetic process
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
phosphatidylinositol 3-kinase signaling
regulation of phosphatidylinositol 3-kinase signaling
viral process
platelet activation
B cell differentiation
positive regulation of cell migration
T cell costimulation
positive regulation of tumor necrosis factor production
cellular response to insulin stimulus
positive regulation of RNA splicing
cellular response to UV
response to endoplasmic reticulum stress
phosphatidylinositol-3-phosphate biosynthetic process
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
positive regulation of transcription factor import into nucleus
negative regulation of apoptotic process
regulation of phosphatidylinositol 3-kinase activity
negative regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
phosphatidylinositol phosphorylation
insulin-like growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
protein stabilization
T cell receptor signaling pathway
leukocyte migration
regulation of stress fiber assembly
NFAT protein import into nucleus
growth hormone receptor signaling pathway
positive regulation of establishment of protein localization to plasma membrane
positive regulation of endoplasmic reticulum unfolded protein response
positive regulation of glucose import in response to insulin stimulus
transmembrane receptor protein tyrosine kinase adaptor activity
insulin receptor binding
insulin-like growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
transcription factor binding
1-phosphatidylinositol-3-kinase activity
protein phosphatase binding
phosphatidylinositol 3-kinase regulator activity
phosphatidylinositol 3-kinase regulatory subunit binding
ErbB-3 class receptor binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
1-phosphatidylinositol-3-kinase regulator activity
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0