Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ADORA1
Transcript:
Assay Name:
Assay ID
CRISPR1127953_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
temperature homeostasis
response to hypoxia
adenosine receptor signaling pathway
regulation of respiratory gaseous exchange by neurological system process
negative regulation of acute inflammatory response
negative regulation of leukocyte migration
positive regulation of peptide secretion
regulation of glomerular filtration
protein targeting to membrane
phagocytosis
inflammatory response
signal transduction
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
cell-cell signaling
nervous system development
negative regulation of cell proliferation
negative regulation of glutamate secretion
lipid catabolic process
negative regulation of synaptic transmission, GABAergic
positive regulation of nucleoside transport
negative regulation of neurotrophin production
positive regulation of protein dephosphorylation
negative regulation of renal sodium excretion
negative regulation of circadian sleep/wake cycle, non-REM sleep
negative regulation of apoptotic process
positive regulation of potassium ion transport
positive regulation of epidermal growth factor-activated receptor activity
negative regulation of blood pressure
positive regulation of blood pressure
negative regulation of vasodilation
negative regulation of hormone secretion
cognition
detection of temperature stimulus involved in sensory perception of pain
negative regulation of lipid catabolic process
regulation of sensory perception of pain
negative regulation of synaptic transmission, glutamatergic
negative regulation of cardiac muscle contraction
excitatory postsynaptic potential
relaxation of vascular smooth muscle
negative regulation of mucus secretion
apoptotic signaling pathway
negative regulation of long term synaptic depression
G-protein coupled adenosine receptor activity
G-protein coupled receptor binding
purine nucleoside binding
phospholipase C activity
protein binding
heat shock protein binding
G-protein beta/gamma-subunit complex binding
heterotrimeric G-protein binding
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0