Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Adora2a
Transcript:
Assay Name:
Assay ID
CRISPR29696_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
synaptic transmission, dopaminergic
adenosine receptor signaling pathway
response to amphetamine
regulation of transcription, DNA-templated
negative regulation of protein kinase activity
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
synaptic transmission, cholinergic
locomotory behavior
negative regulation of cell proliferation
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
positive regulation of glutamate secretion
positive regulation of acetylcholine secretion, neurotransmission
regulation of norepinephrine secretion
positive regulation of cAMP biosynthetic process
response to caffeine
positive regulation of synaptic transmission, GABAergic
synaptic transmission, glutamatergic
positive regulation of urine volume
positive regulation of renal sodium excretion
negative regulation of locomotion
vasodilation
eating behavior
negative regulation of vascular permeability
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to alkaloid
negative regulation of neuron apoptotic process
positive regulation of circadian sleep/wake cycle, sleep
negative regulation of alpha-beta T cell activation
astrocyte activation
regulation of synaptic plasticity
neuron projection morphogenesis
positive regulation of protein secretion
negative regulation of inflammatory response
regulation of mitochondrial membrane potential
membrane depolarization
regulation of calcium ion transport
positive regulation of synaptic transmission, glutamatergic
excitatory postsynaptic potential
inhibitory postsynaptic potential
prepulse inhibition
positive regulation of apoptotic signaling pathway
G-protein coupled adenosine receptor activity
signal transducer activity
G-protein coupled receptor activity
enzyme binding
type 5 metabotropic glutamate receptor binding
identical protein binding
protein heterodimerization activity
alpha-actinin binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0