Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Acvr1c
Transcript:
Assay Name:
Assay ID
CRISPR307694_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
trophectodermal cell proliferation
response to dietary excess
protein phosphorylation
apoptotic process
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor complex assembly
determination of left/right symmetry
response to glucose
phosphorylation
lipid storage
cell differentiation
apoptotic nuclear changes
response to insulin
activin receptor signaling pathway
nodal signaling pathway
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of insulin secretion
negative regulation of trophoblast cell migration
negative regulation of chorionic trophoblast cell proliferation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
ATP binding
kinase activity
activin receptor activity, type I
transferase activity
transferase activity, transferring phosphorus-containing groups
growth factor binding
nodal binding
SMAD binding
metal ion binding
activin binding
BMP receptor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0