Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Bad
Transcript:
Assay Name:
Assay ID
CRISPR38515_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
release of cytochrome c from mitochondria
glucose catabolic process
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
cell proliferation
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of mitochondrial membrane potential
suppression by virus of host apoptotic process
cytokine-mediated signaling pathway
positive regulation of insulin secretion
positive regulation of glucokinase activity
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucose homeostasis
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
type B pancreatic cell proliferation
positive regulation of B cell differentiation
positive regulation of T cell differentiation
positive regulation of proteolysis
negative regulation of cytolysis
ADP metabolic process
ATP metabolic process
regulation of mitochondrial membrane permeability
pore complex assembly
positive regulation of epithelial cell proliferation
positive regulation of apoptotic process by virus
cellular process regulating host cell cycle in response to virus
cellular response to organic substance
cellular response to nicotine
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
intrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity
positive regulation of neuron death
positive regulation of type B pancreatic cell development
protein binding
phospholipid binding
lipid binding
cysteine-type endopeptidase activator activity involved in apoptotic process
protein kinase binding
protein phosphatase binding
protein phosphatase 2B binding
protein kinase B binding
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0