Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Fgfr3
Transcript:
Assay Name:
Assay ID
CRISPR454442_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
skeletal system development
positive regulation of endothelial cell proliferation
morphogenesis of an epithelium
lens morphogenesis in camera-type eye
protein phosphorylation
apoptotic process
cell-cell signaling
positive regulation of cell proliferation
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of phospholipase activity
oligodendrocyte development
phosphorylation
peptidyl-tyrosine phosphorylation
substantia nigra development
central nervous system myelination
cell differentiation
bone mineralization
forebrain development
positive regulation of protein ubiquitination
somatic stem cell population maintenance
post-anal tail morphogenesis
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of MAPK cascade
positive regulation of neuron apoptotic process
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of cell differentiation
negative regulation of mitotic nuclear division
negative regulation of smoothened signaling pathway
protein autophosphorylation
digestive tract morphogenesis
negative regulation of developmental growth
response to axon injury
negative regulation of astrocyte differentiation
inner ear development
negative regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
cartilage development
inner ear receptor cell differentiation
bone morphogenesis
axonogenesis involved in innervation
alveolar secondary septum development
lens fiber cell development
positive regulation of ERK1 and ERK2 cascade
bone maturation
epithelial cell fate commitment
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
cochlea development
positive regulation of canonical Wnt signaling pathway
fibroblast growth factor receptor apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
fibroblast growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0