Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Tgfbr2
Transcript:
Assay Name:
Assay ID
CRISPR478309_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
vasculogenesis
in utero embryonic development
positive regulation of mesenchymal cell proliferation
lens development in camera-type eye
positive regulation of tolerance induction to self antigen
positive regulation of B cell tolerance induction
positive regulation of T cell tolerance induction
growth plate cartilage development
growth plate cartilage chondrocyte growth
protein phosphorylation
receptor-mediated endocytosis
apoptotic process
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
common-partner SMAD protein phosphorylation
Notch signaling pathway
smoothened signaling pathway
gastrulation
brain development
heart development
negative regulation of cell proliferation
organ morphogenesis
regulation of gene expression
positive regulation of epithelial cell migration
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
cell differentiation
lung development
activation of protein kinase activity
embryonic hemopoiesis
regulation of growth
regulation of cell proliferation
response to drug
myeloid dendritic cell differentiation
positive regulation of angiogenesis
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
positive regulation of NK T cell differentiation
cartilage development
palate development
negative regulation of cardiac muscle cell proliferation
pathway-restricted SMAD protein phosphorylation
lung morphogenesis
bronchus development
bronchus morphogenesis
trachea morphogenesis
trachea formation
mammary gland morphogenesis
lung lobe morphogenesis
response to cholesterol
lens fiber cell apoptotic process
positive regulation of reactive oxygen species metabolic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
receptor activity
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor activity, type II
protein binding
ATP binding
glycosaminoglycan binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
mitogen-activated protein kinase kinase kinase binding
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
SMAD binding
metal ion binding
transforming growth factor beta binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0