Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Ngf
Transcript:
Assay Name:
Assay ID
CRISPR482442_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
neurotrophic factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein tyrosine kinase signaling pathway
cell-cell signaling
peripheral nervous system development
memory
positive regulation of cell proliferation
adult locomotory behavior
extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of peptidase activity
positive regulation of gene expression
negative regulation of endopeptidase activity
positive regulation of neuron projection development
positive regulation of neuron maturation
sensory perception of pain
positive regulation of cell growth
neuron projection development
positive regulation of protein ubiquitination
positive regulation of protein autophosphorylation
intracellular signal transduction
nerve growth factor signaling pathway
negative regulation of neuron apoptotic process
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of axon extension
regulation of neurotransmitter secretion
positive regulation of collateral sprouting
neuron projection morphogenesis
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of release of sequestered calcium ion into cytosol
positive regulation of neurotrophin TRK receptor signaling pathway
neuron apoptotic process
cellular response to acid chemical
extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of stem cell proliferation
negative regulation of type B pancreatic cell apoptotic process
enzyme inhibitor activity
receptor signaling protein activity
receptor binding
nerve growth factor receptor binding
growth factor activity
metalloendopeptidase inhibitor activity
peptidase inhibitor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0