Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
INS
Transcript:
Assay Name:
Assay ID
CRISPR485421_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
negative regulation of acute inflammatory response
glucose metabolic process
regulation of transcription, DNA-templated
regulation of cellular amino acid metabolic process
ER to Golgi vesicle-mediated transport
acute-phase response
G-protein coupled receptor signaling pathway
cell-cell signaling
positive regulation of cell proliferation
insulin receptor signaling pathway
positive regulation of gene expression
positive regulation of phosphatidylinositol 3-kinase signaling
glucose transport
regulation of transmembrane transporter activity
positive regulation of cell growth
positive regulation of cell migration
positive regulation of protein autophosphorylation
activation of protein kinase B activity
positive regulation of cellular protein metabolic process
negative regulation of protein oligomerization
regulation of protein localization
negative regulation of NAD(P)H oxidase activity
wound healing
negative regulation of protein catabolic process
glucose homeostasis
positive regulation of MAPK cascade
cellular protein metabolic process
positive regulation of nitric oxide biosynthetic process
positive regulation of cell differentiation
negative regulation of gluconeogenesis
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
negative regulation of glycogen catabolic process
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
negative regulation of proteolysis
negative regulation of vasodilation
positive regulation of vasodilation
negative regulation of fatty acid metabolic process
positive regulation of glucose import
positive regulation of insulin receptor signaling pathway
alpha-beta T cell activation
positive regulation of lipid biosynthetic process
regulation of protein secretion
negative regulation of protein secretion
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of lipid catabolic process
positive regulation of nitric-oxide synthase activity
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein kinase B signaling
fatty acid homeostasis
negative regulation of respiratory burst involved in inflammatory response
positive regulation of respiratory burst
positive regulation of peptide hormone secretion
positive regulation of brown fat cell differentiation
positive regulation of protein localization to nucleus
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
negative regulation of feeding behavior
protease binding
insulin receptor binding
insulin-like growth factor receptor binding
hormone activity
protein binding
identical protein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0