Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Tgfbr3
Transcript:
Assay Name:
Assay ID
CRISPR495084_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
vasculogenesis
osteoblast differentiation
in utero embryonic development
blastocyst development
epithelial to mesenchymal transition
liver development
blood vessel remodeling
epicardial cell to mesenchymal cell transition
protein complex assembly
protein phosphorylation
immune response
transforming growth factor beta receptor signaling pathway
transforming growth factor beta receptor complex assembly
positive regulation of cell proliferation
embryo development
negative regulation of epithelial cell migration
negative regulation of epithelial to mesenchymal transition
cell growth
BMP signaling pathway
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of BMP signaling pathway
response to follicle-stimulating hormone
response to prostaglandin E
response to luteinizing hormone
intracellular signal transduction
regulation of protein binding
negative regulation of epithelial cell proliferation
regulation of blood vessel size
positive regulation of NF-kappaB transcription factor activity
negative regulation of cellular component movement
ventricular cardiac muscle tissue morphogenesis
palate development
cardiac muscle cell proliferation
definitive hemopoiesis
cardiac epithelial to mesenchymal transition
definitive erythrocyte differentiation
heart trabecula formation
pathway-restricted SMAD protein phosphorylation
coronary vasculature development
coronary vasculature morphogenesis
vasculogenesis involved in coronary vascular morphogenesis
visceral serous pericardium development
negative regulation of apoptotic process involved in morphogenesis
transforming growth factor beta-activated receptor activity
type II transforming growth factor beta receptor binding
transforming growth factor beta receptor binding
protein binding
glycosaminoglycan binding
heparin binding
coreceptor activity
fibroblast growth factor binding
PDZ domain binding
SMAD binding
activin binding
transforming growth factor beta binding
transforming growth factor beta receptor activity, type III
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0