Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Fgf8
Transcript:
Assay Name:
Assay ID
CRISPR497501_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
patterning of blood vessels
branching involved in ureteric bud morphogenesis
organ induction
kidney development
neural plate morphogenesis
heart looping
blood vessel remodeling
heart morphogenesis
outflow tract septum morphogenesis
outflow tract morphogenesis
epithelial to mesenchymal transition involved in endocardial cushion formation
apoptotic process
response to oxidative stress
multicellular organism development
determination of left/right symmetry
heart development
motor neuron axon guidance
mesodermal cell migration
positive regulation of cell proliferation
gonad development
fibroblast growth factor receptor signaling pathway
embryo development ending in birth or egg hatching
positive regulation of gene expression
telencephalon development
pallium development
subpallium development
forebrain dorsal/ventral pattern formation
cell proliferation in forebrain
forebrain neuron development
central nervous system neuron development
signal transduction involved in regulation of gene expression
cell differentiation
lung development
BMP signaling pathway
male genitalia development
thyroid gland development
otic vesicle formation
midbrain-hindbrain boundary development
dorsal/ventral axon guidance
embryonic heart tube development
limb morphogenesis
embryonic hindlimb morphogenesis
aorta morphogenesis
inner ear morphogenesis
regulation of odontogenesis of dentin-containing tooth
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
cell fate commitment
positive regulation of mitotic nuclear division
positive regulation of organ growth
generation of neurons
forebrain morphogenesis
positive chemotaxis
positive regulation of cell division
negative regulation of cardiac muscle tissue development
pharyngeal system development
canonical Wnt signaling pathway
corticotropin hormone secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
bone development
lung morphogenesis
branching involved in salivary gland morphogenesis
neuroepithelial cell differentiation
dopaminergic neuron differentiation
cell migration involved in mesendoderm migration
receptor binding
fibroblast growth factor receptor binding
type 1 fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
growth factor activity
chemoattractant activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0