Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FZD5
Transcript:
Assay Name:
Assay ID
CRISPR498687_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
embryonic axis specification
angiogenesis
positive regulation of T cell cytokine production
G-protein coupled receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
synapse assembly
negative regulation of cell proliferation
anterior/posterior axis specification, embryo
neuron differentiation
post-embryonic camera-type eye development
positive regulation of interferon-gamma production
T cell differentiation in thymus
non-canonical Wnt signaling pathway
positive regulation of JUN kinase activity
Wnt signaling pathway involved in dorsal/ventral axis specification
positive regulation of transcription from RNA polymerase II promoter
cell maturation
embryonic camera-type eye morphogenesis
Spemann organizer formation
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
apoptotic process involved in morphogenesis
branching involved in labyrinthine layer morphogenesis
syncytiotrophoblast cell differentiation involved in labyrinthine layer development
labyrinthine layer blood vessel development
chorionic trophoblast cell differentiation
regulation of canonical Wnt signaling pathway
cellular response to molecule of bacterial origin
regulation of chorionic trophoblast cell proliferation
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
beta-catenin destruction complex disassembly
regulation of bicellular tight junction assembly
beta-amyloid binding
G-protein coupled receptor activity
protein binding
Wnt-protein binding
protein kinase binding
ubiquitin protein ligase binding
Wnt-activated receptor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0