Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Tgfb2
Transcript:
Assay Name:
Assay ID
CRISPR513978_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
skeletal system development
cartilage condensation
blood vessel development
eye development
response to hypoxia
epithelial to mesenchymal transition
hair follicle development
blood vessel remodeling
heart morphogenesis
outflow tract morphogenesis
heart valve morphogenesis
cardiac left ventricle morphogenesis
cardiac right ventricle morphogenesis
protein phosphorylation
cell cycle arrest
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
axon guidance
heart development
cell death
positive regulation of cell proliferation
negative regulation of cell proliferation
glial cell migration
cardioblast differentiation
positive regulation of gene expression
positive regulation of epithelial cell migration
negative regulation of alkaline phosphatase activity
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
positive regulation of phosphatidylinositol 3-kinase signaling
cell growth
cell migration
signal transduction by protein phosphorylation
hemopoiesis
extracellular matrix organization
collagen fibril organization
positive regulation of cell growth
negative regulation of cell growth
positive regulation of cell migration
hair follicle morphogenesis
activation of protein kinase activity
response to progesterone
positive regulation of stress-activated MAPK cascade
regulation of transforming growth factor beta2 production
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
growth
wound healing
regulation of cell proliferation
dopamine biosynthetic process
response to drug
catagen
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
regulation of MAPK cascade
positive regulation of neuron apoptotic process
response to estrogen
cell-cell junction organization
negative regulation of keratinocyte differentiation
positive regulation of integrin biosynthetic process
positive regulation of cell cycle
positive regulation of heart contraction
somatic stem cell division
neuron fate commitment
neuron development
embryonic neurocranium morphogenesis
negative regulation of epithelial cell proliferation
positive regulation of protein secretion
negative regulation of immune response
negative regulation of release of sequestered calcium ion into cytosol
positive regulation of cell division
positive regulation of catagen
positive regulation of cardioblast differentiation
cardiac muscle cell proliferation
cardiac epithelial to mesenchymal transition
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
ventricular septum morphogenesis
negative regulation of cartilage development
positive regulation of activation-induced cell death of T cells
extrinsic apoptotic signaling pathway
regulation of extracellular matrix organization
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
beta-amyloid binding
receptor signaling protein serine/threonine kinase activity
receptor binding
type II transforming growth factor beta receptor binding
cytokine activity
transforming growth factor beta receptor binding
protein binding
growth factor activity
type III transforming growth factor beta receptor binding
protein homodimerization activity
protein heterodimerization activity
protein N-terminus binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0