Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Nkx3-1
Transcript:
Assay Name:
Assay ID
CRISPR516929_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
urogenital system development
positive regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
activation of cysteine-type endopeptidase activity involved in apoptotic process
multicellular organism development
salivary gland development
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell death
positive regulation of phosphatidylinositol 3-kinase signaling
androgen receptor signaling pathway
prostate gland development
response to testosterone
internal genitalia morphogenesis
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
steroid hormone mediated signaling pathway
protein kinase B signaling
negative regulation of insulin-like growth factor receptor signaling pathway
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
negative regulation of mitotic cell cycle
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
branching morphogenesis of an epithelial tube
negative regulation of epithelial cell proliferation
positive regulation of cell division
branching involved in prostate gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to steroid hormone stimulus
cellular response to hypoxia
mitotic cell cycle arrest
negative regulation of estrogen receptor binding
positive regulation of androgen secretion
positive regulation of response to DNA damage stimulus
positive regulation of apoptotic signaling pathway
positive regulation of intrinsic apoptotic signaling pathway
transcription regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
core promoter binding
DNA binding
transcription factor activity, sequence-specific DNA binding
androgen receptor activity
protein binding
transcription factor binding
cysteine-type endopeptidase activator activity involved in apoptotic process
estrogen receptor activity
protein kinase activator activity
estrogen receptor binding
histone deacetylase binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
MADS box domain binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0