Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Pik3ca
Transcript:
Assay Name:
Assay ID
CRISPR517027_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
liver development
adaptive immune response
glucose metabolic process
protein phosphorylation
inflammatory response
phosphatidylinositol 3-kinase signaling
phosphorylation
platelet activation
positive regulation of peptidyl-serine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of cellular respiration
protein kinase B signaling
negative regulation of neuron apoptotic process
hypomethylation of CpG island
innate immune response
positive regulation of protein kinase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
cell chemotaxis
adipose tissue development
cellular response to glucose stimulus
energy homeostasis
mitophagy in response to mitochondrial depolarization
negative regulation of fibroblast apoptotic process
regulation of genetic imprinting
negative regulation of anoikis
nucleotide binding
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
1-phosphatidylinositol-3-kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphotransferase activity, alcohol group as acceptor
protein kinase activator activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
insulin receptor substrate binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0