Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Il2
Transcript:
Assay Name:
Assay ID
CRISPR538138_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
interleukin superfamily
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
immune system process
negative regulation of B cell apoptotic process
immune response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
protein kinase C-activating G-protein coupled receptor signaling pathway
positive regulation of B cell proliferation
positive regulation of interferon-gamma production
positive regulation of interleukin-17 production
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
positive regulation of tyrosine phosphorylation of Stat5 protein
positive regulation of T cell differentiation
positive regulation of regulatory T cell differentiation
negative regulation of heart contraction
positive regulation of transcription from RNA polymerase II promoter
regulation of T cell homeostatic proliferation
positive regulation of isotype switching to IgG isotypes
negative regulation of lymphocyte proliferation
negative regulation of inflammatory response
positive regulation of immunoglobulin secretion
positive regulation of dendritic spine development
extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
interleukin-2 receptor binding
growth factor activity
carbohydrate binding
kappa-type opioid receptor binding
glycosphingolipid binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0