Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Mapk8
Transcript:
Assay Name:
Assay ID
CRISPR557637_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
ossification
regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
response to osmotic stress
signal transduction
JNK cascade
JUN phosphorylation
response to heat
response to UV
regulation of gene expression
positive regulation of gene expression
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
positive regulation of cell migration
regulation of histone deacetylation
positive regulation of microtubule polymerization
neuron projection development
positive regulation of cyclase activity
negative regulation of protein binding
regulation of protein localization
response to hydrogen peroxide
regulation of circadian rhythm
negative regulation of apoptotic process
positive regulation of DNA replication
response to cadmium ion
determination of dorsal identity
rhythmic process
positive regulation of protein metabolic process
cellular response to hydrogen peroxide
cellular response to lipopolysaccharide
cellular response to nitric oxide
positive regulation of deacetylase activity
type B pancreatic cell apoptotic process
neuronal stem cell population maintenance
programmed necrotic cell death
positive regulation of determination of dorsal identity
positive regulation of apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
JUN kinase activity
MAP kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
enzyme binding
histone deacetylase regulator activity
histone deacetylase binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0