Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Mapk1
Transcript:
Assay Name:
Assay ID
CRISPR561722_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
MAPK import into nucleus
transcription, DNA-templated
protein phosphorylation
apoptotic process
cellular response to DNA damage stimulus
cell cycle
signal transduction
response to toxic substance
organ morphogenesis
positive regulation of peptidyl-threonine phosphorylation
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
sensory perception of pain
cytosine metabolic process
lipopolysaccharide-mediated signaling pathway
response to lipopolysaccharide
regulation of stress-activated MAPK cascade
mammary gland epithelial cell proliferation
intracellular signal transduction
ERBB signaling pathway
response to exogenous dsRNA
response to estrogen
negative regulation of cell differentiation
positive regulation of translation
T cell receptor signaling pathway
B cell receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
regulation of cytoskeleton organization
long-term synaptic potentiation
lung morphogenesis
trachea formation
labyrinthine layer blood vessel development
ERK1 and ERK2 cascade
response to epidermal growth factor
cellular response to organic substance
caveolin-mediated endocytosis
regulation of Golgi inheritance
cellular response to granulocyte macrophage colony-stimulating factor stimulus
regulation of early endosome to late endosome transport
nucleotide binding
phosphotyrosine binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding
transcription factor binding
RNA polymerase II carboxy-terminal domain kinase activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase kinase binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0