Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Igf1r
Transcript:
Assay Name:
Assay ID
CRISPR562056_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
immune response
transmembrane receptor protein tyrosine kinase signaling pathway
axonogenesis
brain development
epidermis development
organ morphogenesis
negative regulation of muscle cell apoptotic process
phosphorylation
establishment of cell polarity
male sex determination
positive regulation of cell migration
mammary gland development
exocrine pancreas development
neuron projection development
positive regulation of cytokinesis
peptidyl-tyrosine autophosphorylation
negative regulation of apoptotic process
negative regulation of MAPK cascade
positive regulation of MAPK cascade
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of DNA replication
positive regulation of mitotic nuclear division
regulation of JNK cascade
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of DNA metabolic process
protein tetramerization
protein heterooligomerization
inactivation of MAPKK activity
positive regulation of protein kinase B signaling
negative regulation of protein kinase B signaling
prostate gland epithelium morphogenesis
positive regulation of steroid hormone biosynthetic process
nucleotide binding
G-protein alpha-subunit binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
insulin-like growth factor-activated receptor activity
insulin receptor binding
protein binding
insulin-like growth factor binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
insulin-like growth factor I binding
identical protein binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0