Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Ccl5
Transcript:
Assay Name:
Assay ID
CRISPR591586_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
chemokine
cytokine
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of defense response to virus by host
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
inflammatory response
immune response
leukocyte cell-cell adhesion
G-protein coupled receptor signaling pathway
cell-cell signaling
response to toxic substance
positive regulation of gene expression
positive regulation of macrophage chemotaxis
positive regulation of T cell chemotaxis
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of smooth muscle cell migration
positive regulation of cell migration
pseudopodium assembly
positive regulation of cellular biosynthetic process
activation of phospholipase D activity
positive regulation of fever generation
lipopolysaccharide-mediated signaling pathway
positive regulation of cell-cell adhesion mediated by integrin
response to cytokine
positive regulation of homotypic cell-cell adhesion
response to interferon-gamma
response to tumor necrosis factor
T-helper 1 cell diapedesis
positive regulation of T cell proliferation
neutrophil activation
positive regulation of phosphorylation
positive regulation of tyrosine phosphorylation of STAT protein
protein kinase B signaling
cellular protein complex assembly
negative regulation by host of viral transcription
negative regulation of viral genome replication
positive regulation of neuron differentiation
positive regulation of osteoclast differentiation
negative regulation of G-protein coupled receptor protein signaling pathway
positive regulation of angiogenesis
positive regulation of cell adhesion
eosinophil chemotaxis
lymphocyte chemotaxis
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
regulation of insulin secretion
regulation of T cell activation
positive chemotaxis
protein tetramerization
positive regulation of calcium ion transport
negative regulation of cell death
positive regulation of mast cell chemotaxis
positive regulation of protein tyrosine kinase activity
chemokine-mediated signaling pathway
negative regulation of chemokine-mediated signaling pathway
negative regulation of T cell apoptotic process
positive regulation of T cell apoptotic process
cellular response to organic cyclic compound
positive regulation of monocyte chemotaxis
extracellular negative regulation of signal transduction
regulation of neuron death
negative regulation of neuron death
negative regulation of macrophage apoptotic process
negative regulation of receptor activity
positive regulation of chemokine (C-X-C motif) ligand 2 production
positive regulation of T cell migration
positive regulation of natural killer cell chemotaxis
phosphatidylinositol phospholipase C activity
protein kinase activity
cytokine activity
chemokine activity
heparin binding
phospholipase activator activity
receptor signaling protein tyrosine kinase activator activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
chemokine receptor binding
protein homodimerization activity
protein self-association
chemokine receptor antagonist activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0