Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FGF1
Transcript:
Assay Name:
Assay ID
CRISPR592899_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
angiogenesis
organ induction
signal transduction
multicellular organism development
cell proliferation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
anatomical structure morphogenesis
positive regulation of endothelial cell migration
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cell differentiation
lung development
positive regulation of cell migration
activation of protein kinase B activity
cellular response to heat
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of MAP kinase activity
positive regulation of GTPase activity
positive regulation of cholesterol biosynthetic process
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of epithelial cell proliferation
positive regulation of cell division
branch elongation involved in ureteric bud branching
positive regulation of ERK1 and ERK2 cascade
mesonephric epithelium development
regulation of endothelial tube morphogenesis
positive regulation of intracellular signal transduction
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
integrin binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
Hsp70 protein binding
S100 protein binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0