Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
NTRK1
Transcript:
Assay Name:
Assay ID
CRISPR594963_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
positive regulation of protein phosphorylation
protein phosphorylation
microtubule-based movement
axon guidance
aging
learning or memory
circadian rhythm
negative regulation of cell proliferation
response to radiation
programmed cell death involved in cell development
positive regulation of neuron projection development
response to activity
peptidyl-tyrosine phosphorylation
olfactory nerve development
B cell differentiation
response to nutrient levels
nerve growth factor signaling pathway
mechanoreceptor differentiation
response to drug
positive regulation of programmed cell death
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
response to ethanol
positive regulation of Ras protein signal transduction
protein autophosphorylation
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
phosphatidylinositol-mediated signaling
sympathetic nervous system development
response to axon injury
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
positive regulation of NF-kappaB transcription factor activity
response to hydrostatic pressure
response to electrical stimulus
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
Sertoli cell development
axonogenesis involved in innervation
behavioral response to formalin induced pain
positive regulation of ERK1 and ERK2 cascade
cellular response to nicotine
cellular response to nerve growth factor stimulus
transmembrane receptor protein tyrosine kinase activity
GPI-linked ephrin receptor activity
neurotrophin p75 receptor binding
protein binding
ATP binding
nerve growth factor receptor activity
kinase binding
protein homodimerization activity
neurotrophin binding
nerve growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0