Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TRPV4
Transcript:
Assay Name:
Assay ID
CRISPR601832_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
diet induced thermogenesis
calcium ion transport
cellular calcium ion homeostasis
cell volume homeostasis
actin filament organization
cell-cell junction assembly
positive regulation of cytosolic calcium ion concentration
osmosensory signaling pathway
response to mechanical stimulus
positive regulation of gene expression
positive regulation of macrophage chemotaxis
negative regulation of neuron projection development
vasopressin secretion
positive regulation of microtubule depolymerization
actin cytoskeleton reorganization
response to insulin
cellular response to heat
hyperosmotic salinity response
glucose homeostasis
positive regulation of vascular permeability
cortical microtubule organization
positive regulation of JNK cascade
microtubule polymerization
regulation of response to osmotic stress
positive regulation of inflammatory response
multicellular organismal water homeostasis
positive regulation of ERK1 and ERK2 cascade
calcium ion import
calcium ion transmembrane transport
cellular response to osmotic stress
cellular hypotonic response
cellular hypotonic salinity response
positive regulation of monocyte chemotactic protein-1 production
positive regulation of macrophage inflammatory protein 1 alpha production
positive regulation of chemokine (C-C motif) ligand 5 production
blood vessel endothelial cell delamination
negative regulation of brown fat cell differentiation
signal transduction involved in regulation of aerobic respiration
positive regulation of chemokine (C-X-C motif) ligand 1 production
positive regulation of energy homeostasis
positive regulation of interleukin-6 secretion
actin binding
osmosensor activity
protein kinase C binding
cation channel activity
calcium channel activity
protein binding
calmodulin binding
ATP binding
microtubule binding
stretch-activated, cation-selective, calcium channel activity
protein kinase binding
SH2 domain binding
alpha-tubulin binding
beta-tubulin binding
actin filament binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0