Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HGF
Transcript:
Assay Name:
Assay ID
CRISPR606578_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
annexin
calcium-binding protein
calmodulin
enzyme modulator
hydrolase
intracellular calcium-sensing protein
peptide hormone
protease
protease inhibitor
receptor
serine protease
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
epithelial to mesenchymal transition
liver development
platelet degranulation
proteolysis
mitotic nuclear division
positive regulation of cell proliferation
negative regulation of autophagy
positive regulation of phosphatidylinositol 3-kinase signaling
hyaluronan metabolic process
positive regulation of cell migration
organ regeneration
positive regulation of myelination
negative regulation of interleukin-6 production
positive regulation of interleukin-10 production
negative regulation of peptidyl-serine phosphorylation
cellular response to hepatocyte growth factor stimulus
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of osteoblast differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
hepatocyte growth factor receptor signaling pathway
negative regulation of inflammatory response
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
myoblast proliferation
cell chemotaxis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
positive regulation of neuron projection regeneration
negative regulation of release of cytochrome c from mitochondria
regulation of p38MAPK cascade
negative regulation of hydrogen peroxide-mediated programmed cell death
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of tau-protein kinase activity
positive regulation of DNA biosynthetic process
serine-type endopeptidase activity
protein binding
growth factor activity
chemoattractant activity
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0