Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
HRAS
Transcript:
Assay Name:
Assay ID
CRISPR608026_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein
enzyme modulator
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of protein phosphorylation
stimulatory C-type lectin receptor signaling pathway
endocytosis
chemotaxis
cell cycle arrest
mitotic cell cycle checkpoint
signal transduction
cell surface receptor signaling pathway
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
axon guidance
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
organ morphogenesis
negative regulation of gene expression
positive regulation of cell migration
positive regulation of interferon-gamma production
negative regulation of GTPase activity
social behavior
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
T-helper 1 type immune response
defense response to protozoan
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
positive regulation of GTPase activity
positive regulation of DNA replication
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
positive regulation of Ras protein signal transduction
ephrin receptor signaling pathway
regulation of long-term neuronal synaptic plasticity
positive regulation of epithelial cell proliferation
T cell receptor signaling pathway
leukocyte migration
protein heterooligomerization
positive regulation of ERK1 and ERK2 cascade
positive regulation of wound healing
cellular senescence
intrinsic apoptotic signaling pathway
positive regulation of ruffle assembly
positive regulation of actin cytoskeleton reorganization
positive regulation of miRNA metabolic process
GTPase activity
protein binding
GTP binding
protein C-terminus binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0