Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CRHR2
Transcript:
Assay Name:
Assay ID
CRISPR616060_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
cell surface receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
hormone-mediated signaling pathway
positive regulation of heart rate
positive regulation of gene expression
negative regulation of gene expression
negative regulation of norepinephrine secretion
positive regulation of serotonin secretion
sensory perception of pain
cerebellum development
hypothalamus development
negative regulation of cAMP biosynthetic process
positive regulation of cAMP biosynthetic process
epithelial cell differentiation
positive regulation of interleukin-6 production
negative regulation of epinephrine secretion
positive regulation of stress-activated MAPK cascade
negative regulation of luteinizing hormone secretion
gastric motility
catecholamine biosynthetic process
positive regulation of blood pressure
negative regulation of follicle-stimulating hormone secretion
skeletal muscle tissue growth
positive regulation of NF-kappaB transcription factor activity
long-term synaptic potentiation
negative regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of ERK1 and ERK2 cascade
cellular response to corticotropin-releasing hormone stimulus
cellular response to glucocorticoid stimulus
negative regulation of calcium ion import
negative regulation of feeding behavior
negative regulation of defecation
protein binding
corticotrophin-releasing factor receptor activity
peptide hormone binding
corticotropin-releasing hormone receptor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0