Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GHRHR
Transcript:
Assay Name:
Assay ID
CRISPR645913_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
lactation
positive regulation of cell proliferation
determination of adult lifespan
cAMP-mediated signaling
water homeostasis
growth hormone secretion
positive regulation of cAMP biosynthetic process
response to insulin
cellular response to insulin stimulus
regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of multicellular organism growth
hormone metabolic process
positive regulation of insulin-like growth factor receptor signaling pathway
response to estrogen
positive regulation of circadian sleep/wake cycle, non-REM sleep
cell maturation
multicellular organismal reproductive process
regulation of protein metabolic process
response to glucocorticoid
positive regulation of growth hormone secretion
somatotropin secreting cell development
cellular response to glucose stimulus
G-protein coupled receptor activity
protein binding
growth hormone-releasing hormone receptor activity
peptide hormone binding
growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0