Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
GPER1
Transcript:
Assay Name:
Assay ID
CRISPR656355_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
positive regulation of neurotransmitter secretion
negative regulation of leukocyte activation
inflammatory response
cell cycle
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
positive regulation of gene expression
negative regulation of gene expression
negative regulation of cell cycle process
positive regulation of phosphatidylinositol 3-kinase signaling
neuronal action potential
apoptotic chromosome condensation
nuclear fragmentation involved in apoptotic nuclear change
positive regulation of cell migration
intracellular steroid hormone receptor signaling pathway
positive regulation of cAMP biosynthetic process
mineralocorticoid receptor signaling pathway
positive regulation of insulin secretion
positive regulation of inositol trisphosphate biosynthetic process
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
steroid hormone mediated signaling pathway
positive regulation of MAPK cascade
innate immune response
negative regulation of fat cell differentiation
positive regulation of epidermal growth factor receptor signaling pathway
positive regulation of G-protein coupled receptor protein signaling pathway
positive regulation of vasodilation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of inflammatory response
positive regulation of neurogenesis
negative regulation of DNA metabolic process
negative regulation of lipid biosynthetic process
positive regulation of release of sequestered calcium ion into cytosol
regulation of cytosolic calcium ion concentration
negative regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of uterine smooth muscle contraction
negative regulation of cell cycle arrest
cellular response to glucose stimulus
cellular response to tumor necrosis factor
cellular response to peptide hormone stimulus
cellular response to mineralocorticoid stimulus
cellular response to estradiol stimulus
positive regulation of establishment of protein localization to plasma membrane
positive regulation of release of cytochrome c from mitochondria
negative regulation of vascular smooth muscle cell proliferation
positive regulation of endothelial cell apoptotic process
positive regulation of cardiac vascular smooth muscle cell differentiation
positive regulation of extrinsic apoptotic signaling pathway
chromatin binding
G-protein coupled receptor activity
steroid binding
protein binding
drug binding
mineralocorticoid receptor activity
estrogen receptor activity
steroid hormone binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0