Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TGFBR1
Transcript:
Assay Name:
Assay ID
CRISPR685965_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
skeletal system development
angiogenesis
in utero embryonic development
kidney development
blastocyst development
epithelial to mesenchymal transition
negative regulation of endothelial cell proliferation
positive regulation of endothelial cell proliferation
lens development in camera-type eye
ventricular trabecula myocardium morphogenesis
ventricular compact myocardium morphogenesis
proepicardium development
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
cell cycle arrest
signal transduction
transforming growth factor beta receptor signaling pathway
heart development
positive regulation of cell proliferation
germ cell migration
male gonad development
post-embryonic development
anterior/posterior pattern specification
regulation of epithelial to mesenchymal transition
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
collagen fibril organization
positive regulation of cell growth
positive regulation of cell migration
negative regulation of transforming growth factor beta receptor signaling pathway
regulation of protein ubiquitination
negative regulation of chondrocyte differentiation
activin receptor signaling pathway
intracellular signal transduction
wound healing
endothelial cell activation
extracellular structure organization
regulation of protein binding
endothelial cell migration
positive regulation of transcription, DNA-templated
thymus development
neuron fate commitment
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
mesenchymal cell differentiation
artery morphogenesis
cell motility
positive regulation of cellular component movement
positive regulation of filopodium assembly
positive regulation of stress fiber assembly
positive regulation of protein kinase B signaling
parathyroid gland development
palate development
pharyngeal system development
regulation of cardiac muscle cell proliferation
cardiac epithelial to mesenchymal transition
pathway-restricted SMAD protein phosphorylation
positive regulation of SMAD protein import into nucleus
ventricular septum morphogenesis
coronary artery morphogenesis
response to cholesterol
cellular response to transforming growth factor beta stimulus
positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of occluding junction disassembly
positive regulation of apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta-activated receptor activity
transforming growth factor beta receptor activity, type I
receptor signaling protein activity
type II transforming growth factor beta receptor binding
protein binding
ATP binding
growth factor binding
SMAD binding
metal ion binding
transforming growth factor beta binding
I-SMAD binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0