Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
AKT1
Transcript:
Assay Name:
Assay ID
CRISPR726508_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
osteoblast differentiation
maternal placenta development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
glycogen biosynthetic process
regulation of glycogen biosynthetic process
glucose metabolic process
translation
regulation of translation
cellular protein modification process
protein phosphorylation
negative regulation of protein kinase activity
nitric oxide biosynthetic process
activation-induced cell death of T cells
inflammatory response
cellular response to DNA damage stimulus
response to oxidative stress
signal transduction
G-protein coupled receptor signaling pathway
germ cell development
aging
cell proliferation
insulin receptor signaling pathway
apoptotic mitochondrial changes
response to heat
negative regulation of autophagy
negative regulation of gene expression
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of fibroblast migration
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
negative regulation of endopeptidase activity
regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
glucose transport
phosphorylation
protein ubiquitination
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
spinal cord development
cell projection organization
cell differentiation
protein catabolic process
platelet activation
hyaluronan metabolic process
positive regulation of cell growth
regulation of cell migration
endocrine pancreas development
T cell costimulation
regulation of myelination
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
lipopolysaccharide-mediated signaling pathway
negative regulation of fatty acid beta-oxidation
positive regulation of endodeoxyribonuclease activity
response to food
positive regulation of cellular protein metabolic process
peripheral nervous system myelin maintenance
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to insulin stimulus
positive regulation of peptidyl-serine phosphorylation
response to fluid shear stress
intracellular signal transduction
cellular response to vascular endothelial growth factor stimulus
ERBB2 signaling pathway
glucose homeostasis
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of mRNA stability
protein kinase B signaling
positive regulation of blood vessel endothelial cell migration
positive regulation of nitric oxide biosynthetic process
positive regulation of fat cell differentiation
positive regulation of glycogen biosynthetic process
positive regulation of epidermal growth factor receptor signaling pathway
negative regulation of cell size
negative regulation of proteolysis
positive regulation of vasoconstriction
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
negative regulation of JNK cascade
protein autophosphorylation
positive regulation of lipid biosynthetic process
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
regulation of nitric-oxide synthase activity
positive regulation of nitric-oxide synthase activity
positive regulation of sequence-specific DNA binding transcription factor activity
striated muscle cell differentiation
response to growth hormone
mammary gland epithelial cell differentiation
glycogen cell differentiation involved in embryonic placenta development
labyrinthine layer blood vessel development
response to UV-A
cellular response to mechanical stimulus
cellular response to epidermal growth factor stimulus
cellular response to prostaglandin E stimulus
cellular response to organic cyclic compound
cellular response to hypoxia
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of establishment of protein localization to plasma membrane
negative regulation of release of cytochrome c from mitochondria
cellular response to granulocyte macrophage colony-stimulating factor stimulus
execution phase of apoptosis
chemical synaptic transmission, postsynaptic
negative regulation of protein kinase activity by protein phosphorylation
positive regulation of protein localization to nucleus
negative regulation of neuron death
regulation of signal transduction by p53 class mediator
regulation of cell cycle checkpoint
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
cellular response to nerve growth factor stimulus
response to insulin-like growth factor stimulus
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein kinase C binding
protein binding
ATP binding
phosphatidylinositol-3,4,5-trisphosphate binding
kinase activity
enzyme binding
nitric-oxide synthase regulator activity
GTPase activating protein binding
identical protein binding
phosphatidylinositol-3,4-bisphosphate binding
protein phosphatase 2A binding
14-3-3 protein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0