Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PDGFRB
Transcript:
Assay Name:
Assay ID
CRISPR732946_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
glycosaminoglycan biosynthetic process
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of cell proliferation
response to toxic substance
positive regulation of phospholipase C activity
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of smooth muscle cell migration
cell migration
peptidyl-tyrosine phosphorylation
positive regulation of cell migration
response to estradiol
positive regulation of phosphoprotein phosphatase activity
response to retinoic acid
regulation of actin cytoskeleton organization
positive regulation of collagen biosynthetic process
response to fluid shear stress
cell migration involved in vasculogenesis
metanephric mesenchymal cell migration
platelet-derived growth factor receptor-beta signaling pathway
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
aorta morphogenesis
cellular response to platelet-derived growth factor stimulus
positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
wound healing
response to hydrogen peroxide
negative regulation of apoptotic process
positive regulation of MAP kinase activity
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
positive regulation of mitotic nuclear division
phosphatidylinositol metabolic process
protein autophosphorylation
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of smooth muscle cell proliferation
inner ear development
positive regulation of chemotaxis
cardiac myofibril assembly
response to hyperoxia
cell chemotaxis
cell migration involved in coronary angiogenesis
retina vasculature development in camera-type eye
positive regulation of ERK1 and ERK2 cascade
smooth muscle cell chemotaxis
metanephric mesenchyme development
metanephric glomerular mesangial cell proliferation involved in metanephros development
metanephric glomerular capillary formation
metanephric comma-shaped body morphogenesis
metanephric S-shaped body morphogenesis
positive regulation of calcium ion import
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
protein tyrosine kinase activity
platelet activating factor receptor activity
platelet-derived growth factor-activated receptor activity
platelet-derived growth factor beta-receptor activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
platelet-derived growth factor receptor binding
protein binding
ATP binding
protein kinase binding
vascular endothelial growth factor binding
phosphatidylinositol 3-kinase binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
platelet-derived growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0