Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PRMT5
Transcript:
Assay Name:
Assay ID
CRISPR752746_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal snRNP assembly
DNA-templated transcription, termination
regulation of transcription, DNA-templated
regulation of mitotic nuclear division
cell proliferation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
circadian regulation of gene expression
peptidyl-arginine N-methylation
endothelial cell activation
histone H4-R3 methylation
regulation of DNA methylation
negative regulation of cell differentiation
positive regulation of oligodendrocyte differentiation
regulation of ERK1 and ERK2 cascade
Golgi ribbon formation
liver regeneration
regulation of signal transduction by p53 class mediator
negative regulation of nucleic acid-templated transcription
positive regulation of adenylate cyclase-inhibiting dopamine receptor signaling pathway
core promoter sequence-specific DNA binding
transcription corepressor activity
protein binding
methyltransferase activity
methyl-CpG binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
protein complex binding
protein-arginine omega-N symmetric methyltransferase activity
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0