Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
IL12A
Transcript:
Assay Name:
Assay ID
CRISPR768302_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
interleukin superfamily
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
immune response
cell cycle arrest
response to virus
response to UV-B
cell migration
response to lipopolysaccharide
negative regulation of interleukin-17 production
positive regulation of interferon-gamma production
positive regulation of natural killer cell activation
positive regulation of mononuclear cell proliferation
positive regulation of smooth muscle cell apoptotic process
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat4 protein
defense response to protozoan
positive regulation of T cell differentiation
positive regulation of cell adhesion
positive regulation of natural killer cell mediated cytotoxicity
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
defense response to Gram-positive bacterium
positive regulation of NK T cell activation
cellular response to lipopolysaccharide
extrinsic apoptotic signaling pathway
cellular response to virus
positive regulation of dendritic cell chemotaxis
cytokine activity
interleukin-12 receptor binding
protein binding
growth factor activity
interleukin-12 beta subunit binding
interleukin-27 binding
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0