Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TGFB2
Transcript:
Assay Name:
Assay ID
CRISPR803250_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
cartilage condensation
angiogenesis
eye development
response to hypoxia
epithelial to mesenchymal transition
hair follicle development
blood vessel remodeling
platelet degranulation
heart morphogenesis
outflow tract septum morphogenesis
membranous septum morphogenesis
heart valve morphogenesis
atrioventricular valve morphogenesis
pulmonary valve morphogenesis
endocardial cushion morphogenesis
cardiac right ventricle morphogenesis
ventricular trabecula myocardium morphogenesis
endocardial cushion fusion
atrial septum primum morphogenesis
protein phosphorylation
cell cycle arrest
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
cell-cell signaling
axon guidance
salivary gland morphogenesis
heart development
cell death
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
glial cell migration
response to wounding
embryo development
cardioblast differentiation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of epithelial cell migration
negative regulation of alkaline phosphatase activity
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
positive regulation of phosphatidylinositol 3-kinase signaling
cell growth
cell migration
signal transduction by protein phosphorylation
hemopoiesis
collagen fibril organization
positive regulation of cell growth
negative regulation of cell growth
neutrophil chemotaxis
hair follicle morphogenesis
activation of protein kinase activity
response to progesterone
positive regulation of stress-activated MAPK cascade
regulation of transforming growth factor beta2 production
regulation of actin cytoskeleton organization
positive regulation of cell adhesion mediated by integrin
ascending aorta morphogenesis
wound healing
dopamine biosynthetic process
odontogenesis
response to drug
catagen
uterine wall breakdown
positive regulation of neuron apoptotic process
cell-cell junction organization
positive regulation of integrin biosynthetic process
positive regulation of ossification
positive regulation of cell cycle
positive regulation of heart contraction
negative regulation of Ras protein signal transduction
somatic stem cell division
embryonic digestive tract development
neuron fate commitment
neuron development
generation of neurons
negative regulation of epithelial cell proliferation
positive regulation of protein secretion
negative regulation of immune response
positive regulation of immune response
positive regulation of cell division
positive regulation of catagen
positive regulation of cardioblast differentiation
cardiac muscle cell proliferation
cardiac epithelial to mesenchymal transition
face morphogenesis
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
ventricular septum morphogenesis
atrial septum morphogenesis
negative regulation of cartilage development
pharyngeal arch artery morphogenesis
positive regulation of activation-induced cell death of T cells
positive regulation of extracellular matrix disassembly
extrinsic apoptotic signaling pathway
positive regulation of protein localization to nucleus
regulation of apoptotic process involved in outflow tract morphogenesis
regulation of complement-dependent cytotoxicity
positive regulation of GTP binding
negative regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
beta-amyloid binding
receptor signaling protein serine/threonine kinase activity
receptor binding
type II transforming growth factor beta receptor binding
cytokine activity
transforming growth factor beta receptor binding
protein binding
growth factor activity
type III transforming growth factor beta receptor binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0