Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
EGFR
Transcript:
Assay Name:
Assay ID
CRISPR813594_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
cell morphogenesis
ossification
embryonic placenta development
positive regulation of protein phosphorylation
hair follicle development
response to stress
signal transduction
cell surface receptor signaling pathway
epidermal growth factor receptor signaling pathway
activation of phospholipase C activity
salivary gland morphogenesis
learning or memory
cell proliferation
positive regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
single organismal cell-cell adhesion
peptidyl-tyrosine phosphorylation
cerebral cortex cell migration
positive regulation of cell growth
positive regulation of cell migration
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
positive regulation of catenin import into nucleus
ERBB2 signaling pathway
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of protein catabolic process
positive regulation of phosphorylation
activation of phospholipase A2 activity by calcium-mediated signaling
negative regulation of apoptotic process
positive regulation of MAP kinase activity
positive regulation of GTPase activity
positive regulation of nitric oxide biosynthetic process
positive regulation of DNA repair
positive regulation of DNA replication
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
digestive tract morphogenesis
positive regulation of epithelial cell proliferation
regulation of peptidyl-tyrosine phosphorylation
regulation of nitric-oxide synthase activity
protein insertion into membrane
positive regulation of protein kinase B signaling
morphogenesis of an epithelial fold
eyelid development in camera-type eye
response to UV-A
positive regulation of ERK1 and ERK2 cascade
cellular response to amino acid stimulus
cellular response to epidermal growth factor stimulus
cellular response to estradiol stimulus
positive regulation of production of miRNAs involved in gene silencing by miRNA
regulation of cell motility
chromatin binding
double-stranded DNA binding
MAP kinase kinase kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
transmembrane signaling receptor activity
epidermal growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
enzyme binding
protein phosphatase binding
nitric-oxide synthase regulator activity
ubiquitin protein ligase binding
identical protein binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
actin filament binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0