Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CDKN1A
Transcript:
Assay Name:
Assay ID
CRISPR865095_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
enzyme modulator
kinase inhibitor
kinase modulator
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
Ras protein signal transduction
negative regulation of cell proliferation
response to toxic substance
response to X-ray
response to organonitrogen compound
negative regulation of gene expression
negative regulation of cell growth
positive regulation of B cell proliferation
organ regeneration
cellular response to extracellular stimulus
regulation of protein import into nucleus, translocation
cellular response to amino acid starvation
cellular response to heat
negative regulation of phosphorylation
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of apoptotic process
positive regulation of programmed cell death
negative regulation of cyclin-dependent protein serine/threonine kinase activity
response to arsenic-containing substance
positive regulation of fibroblast proliferation
protein stabilization
response to corticosterone
response to hyperoxia
intestinal epithelial cell maturation
cellular response to ionizing radiation
cellular response to UV-B
mitotic cell cycle arrest
cellular senescence
replicative senescence
stress-induced premature senescence
intrinsic apoptotic signaling pathway
positive regulation of cyclin-dependent protein kinase activity
negative regulation of G1/S transition of mitotic cell cycle
regulation of DNA biosynthetic process
positive regulation of reactive oxygen species metabolic process
cyclin-dependent protein serine/threonine kinase inhibitor activity
protein binding
cyclin-dependent protein kinase activating kinase activity
cyclin binding
ubiquitin protein ligase binding
protein complex binding
metal ion binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0