Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FOXO1
Transcript:
Assay Name:
Assay ID
CRISPR889863_SGM
Size
1.5 nmol
1.5 nmol
Availability
Made To Order
Catalog #
A35533
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA binding protein
nucleic acid binding
transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
temperature homeostasis
cellular glucose homeostasis
transcription, DNA-templated
protein acetylation
autophagy
apoptotic process
cellular response to DNA damage stimulus
insulin receptor signaling pathway
cellular response to starvation
positive regulation of autophagy
endocrine pancreas development
cellular response to insulin stimulus
negative regulation of stress-activated MAPK cascade
cellular response to oxidative stress
regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
positive regulation of apoptotic process
negative regulation of apoptotic process
fat cell differentiation
negative regulation of fat cell differentiation
positive regulation of gluconeogenesis
positive regulation of protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
enamel mineralization
cellular response to hydrogen peroxide
cellular response to cold
cellular response to hyperoxia
cellular response to dexamethasone stimulus
cellular response to nitric oxide
negative regulation of canonical Wnt signaling pathway
neuronal stem cell population maintenance
response to fluoride
regulation of neural precursor cell proliferation
regulation of reactive oxygen species metabolic process
regulation of energy homeostasis
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
chromatin binding
protein binding
beta-catenin binding
ubiquitin protein ligase binding
sequence-specific DNA binding
protein phosphatase 2A binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0