Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
EGF
Transcript:
Assay Name:
Assay ID
CRISPR893662_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
extracellular matrix protein
receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
activation of MAPK activity
angiogenesis
platelet degranulation
DNA replication
signal transduction
activation of transmembrane receptor protein tyrosine kinase activity
epidermal growth factor receptor signaling pathway
STAT protein import into nucleus
positive regulation of cell proliferation
positive regulation of peptidyl-threonine phosphorylation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
positive regulation of cerebellar granule cell precursor proliferation
positive regulation of catenin import into nucleus
ERBB2 signaling pathway
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of phosphorylation
positive regulation of DNA binding
positive regulation of MAP kinase activity
positive regulation of GTPase activity
Wnt signaling pathway involved in dorsal/ventral axis specification
positive regulation of epidermal growth factor-activated receptor activity
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
branching morphogenesis of an epithelial tube
negative regulation of secretion
canonical Wnt signaling pathway
mammary gland alveolus development
ERK1 and ERK2 cascade
regulation of calcium ion import
negative regulation of cholesterol efflux
positive regulation of hyaluronan biosynthetic process
regulation of protein localization to cell surface
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
regulation of cell motility
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
epidermal growth factor receptor binding
calcium ion binding
protein binding
growth factor activity
Wnt-protein binding
transmembrane receptor protein tyrosine kinase activator activity
Wnt-activated receptor activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0