Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
PDGFB
Transcript:
Assay Name:
Assay ID
CRISPR928019_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
response to hypoxia
embryonic placenta development
positive regulation of endothelial cell proliferation
monocyte chemotaxis
platelet degranulation
positive regulation of glomerular filtration
DNA replication
protein phosphorylation
transforming growth factor beta receptor signaling pathway
heart development
positive regulation of cell proliferation
response to wounding
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
positive regulation of gene expression
negative regulation of gene expression
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of smooth muscle cell migration
cell growth
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
hemopoiesis
extracellular matrix organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
negative regulation of protein binding
activation of protein kinase activity
activation of protein kinase B activity
response to estradiol
response to insulin
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
paracrine signaling
wound healing
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of blood vessel endothelial cell migration
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
response to estrogen
positive regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of DNA replication
positive regulation of mitotic nuclear division
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of chemotaxis
positive regulation of cell division
cell chemotaxis
positive regulation of protein tyrosine kinase activity
positive regulation of ERK1 and ERK2 cascade
protein kinase C signaling
cellular response to growth factor stimulus
cellular response to mycophenolic acid
positive regulation of glomerular mesangial cell proliferation
metanephric glomerular mesangial cell development
reactive oxygen species metabolic process
positive regulation of calcium ion import
positive regulation of hyaluronan biosynthetic process
negative regulation of pri-miRNA transcription from RNA polymerase II promoter
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
positive regulation of vascular smooth muscle cell proliferation
positive regulation of vascular associated smooth muscle cell migration
negative regulation of vascular smooth muscle cell differentiation
positive regulation of vascular smooth muscle cell dedifferentiation
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
positive regulation of metanephric mesenchymal cell migration
Ras guanyl-nucleotide exchange factor activity
platelet-derived growth factor receptor binding
protein binding
collagen binding
growth factor activity
superoxide-generating NADPH oxidase activator activity
chemoattractant activity
identical protein binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
platelet-derived growth factor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0