Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
MAPK9
Transcript:
Assay Name:
Assay ID
CRISPR947318_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
release of cytochrome c from mitochondria
positive regulation of protein phosphorylation
protein phosphorylation
protein targeting to mitochondrion
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to stress
JNK cascade
JUN phosphorylation
central nervous system development
response to mechanical stimulus
response to toxic substance
positive regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of cell morphogenesis involved in differentiation
response to amine
neuron projection development
positive regulation of prostaglandin biosynthetic process
regulation of protein ubiquitination
positive regulation of prostaglandin secretion
positive regulation of chemokine production
cellular response to UV
Fc-epsilon receptor signaling pathway
response to drug
regulation of circadian rhythm
positive regulation of nitric oxide biosynthetic process
positive regulation of transcription, DNA-templated
regulation of JNK cascade
response to cadmium ion
rhythmic process
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of nitric-oxide synthase biosynthetic process
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
positive regulation of podosome assembly
positive regulation of apoptotic signaling pathway
JUN kinase activity
protein binding
ATP binding
transcription factor binding
cysteine-type endopeptidase activator activity involved in apoptotic process
mitogen-activated protein kinase kinase kinase binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0