Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
KDR
Transcript:
Assay Name:
Assay ID
CRISPR972749_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
vasculogenesis
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
endothelium development
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cell proliferation
regulation of cell shape
positive regulation of endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
viral process
positive regulation of macroautophagy
peptidyl-tyrosine phosphorylation
signal transduction by protein phosphorylation
extracellular matrix organization
positive regulation of cell migration
embryonic hemopoiesis
calcium-mediated signaling using intracellular calcium source
cellular response to vascular endothelial growth factor stimulus
positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
peptidyl-tyrosine autophosphorylation
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of angiogenesis
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
positive regulation of positive chemotaxis
positive regulation of nitric-oxide synthase biosynthetic process
positive regulation of focal adhesion assembly
positive regulation of mitochondrial depolarization
positive regulation of ERK1 and ERK2 cascade
positive regulation of mitochondrial fission
negative regulation of endothelial cell apoptotic process
positive regulation of vasculogenesis
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
vascular endothelial growth factor-activated receptor activity
integrin binding
protein binding
ATP binding
growth factor binding
Hsp90 protein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0