Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_000142.4 | NP_000133.1 | ||||
NM_001163213.1 | NP_001156685.1 | |||||
NM_022965.3 | NP_075254.1 | |||||
XM_006713868.1 | XP_006713931.1 | |||||
XM_006713869.1 | XP_006713932.1 | |||||
XM_006713870.1 | XP_006713933.1 | |||||
XM_006713871.1 | XP_006713934.1 | |||||
XM_006713872.1 | XP_006713935.1 | |||||
XM_006713873.1 | XP_006713936.1 | |||||
XM_011513420.1 | XP_011511722.1 | |||||
XM_011513422.1 | XP_011511724.1 | |||||
GenBank mRNA | AB209441.1 | - | ||||
AF238374.1 | - | |||||
AF245114.1 | - | |||||
AF369211.1 | - | |||||
AF369212.1 | - | |||||
AF369213.1 | - | |||||
AK308936.1 | - | |||||
AW204106.1 | - | |||||
BC121175.2 | - | |||||
BC128610.1 | - | |||||
BC153824.1 | - | |||||
DR003490.1 | - | |||||
ENST00000440486.0 | - | |||||
HM244683.1 | - | |||||
KJ534834.1 | - | |||||
KJ534974.1 | - | |||||
KJ535004.1 | - | |||||
M58051.1 | - | |||||
M59374.1 | - | |||||
M64347.1 | - | |||||
X84939.1 | - |