Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_002546.3 | NP_002537.3 | ||||
GenBank mRNA | AB002146.1 | - | ||||
AF134187.1 | - | |||||
AK223155.1 | - | |||||
AK308524.1 | - | |||||
AK313710.1 | - | |||||
BC011023.1 | - | |||||
BC030155.2 | - | 4875170 | ||||
BM748946.1 | - | |||||
BQ000897.1 | - | |||||
CK903484.1 | - | |||||
DA477521.1 | - | |||||
ENST00000297350.0 | - | |||||
U94332.1 | - |