Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001322809.1 | NP_001309738.1 | ||||
NM_015985.3 | NP_057069.1 | |||||
XM_011529239.2 | XP_011527541.1 | |||||
GenBank mRNA | AF074332.1 | - | ||||
AF113708.1 | - | |||||
AK304756.1 | - | |||||
BC111976.1 | - | 8327444 | ||||
BC111978.1 | - | 8327446 | ||||
BM999095.1 | - | |||||
ENST00000381922.0 | - |