Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001178130.2 | NP_001171601.1 | ||||
NM_001178131.2 | NP_001171602.1 | |||||
NM_001963.5 | NP_001954.2 | |||||
XM_005262796.2 | XP_005262853.1 | |||||
XM_011531707.1 | XP_011530009.1 | |||||
XM_017007845.1 | XP_016863334.1 | |||||
XM_017007846.1 | XP_016863335.1 | |||||
XM_017007847.1 | XP_016863336.1 | |||||
XM_017007848.1 | XP_016863337.1 | |||||
XM_017007849.1 | XP_016863338.1 | |||||
XM_017007850.1 | XP_016863339.1 | |||||
XM_017007851.1 | XP_016863340.1 | |||||
XM_017007852.1 | XP_016863341.1 | |||||
XM_017007853.1 | XP_016863342.1 | |||||
XM_017007854.1 | XP_016863343.1 | |||||
XM_017007855.1 | XP_016863344.1 | |||||
GenBank mRNA | AI628974.1 | - | ||||
AK299306.1 | - | |||||
AK307734.1 | - | |||||
AY548762.1 | - | |||||
BC093731.1 | - | 7939576 | ||||
BC113461.1 | - | 8322513 | ||||
BC143357.1 | - | |||||
CA437074.1 | - | |||||
CU446298.1 | - | |||||
DB227969.1 | - | |||||
DB349864.1 | - | |||||
ENST00000265171.0 | - | |||||
JQ346088.1 | - | |||||
X04571.1 | - |