Assay Details
Gene Symbol: PIK3CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_006218.3 | NP_006209.2 | ||||
XM_006713658.3 | XP_006713721.1 | |||||
XM_011512894.2 | XP_011511196.1 | |||||
GenBank mRNA | AB210020.1 | - | ||||
AI379480.1 | - | |||||
AK021510.1 | - | |||||
AK292940.1 | - | |||||
AK297203.1 | - | |||||
AK302608.1 | - | |||||
AU129890.2 | - | |||||
AW445128.1 | - | |||||
BC113601.1 | - | 8322653 | ||||
BC113603.1 | - | 8322655 | ||||
BQ228953.1 | - | |||||
BX437219.2 | - | |||||
BX473473.1 | - | |||||
BX484734.1 | - | |||||
DQ910907.1 | - | |||||
HM437241.1 | - | |||||
U79143.1 | - | |||||
Z29090.1 | - |
