Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_005585.4 | NP_005576.3 | ||||
NR_027654.1 | ||||||
XM_011521561.2 | XP_011519863.1 | |||||
GenBank mRNA | AF035528.1 | - | ||||
AF037469.1 | - | |||||
AF043640.1 | - | |||||
AM909653.1 | - | |||||
BC012986.2 | - | 3632931 | ||||
BC029288.1 | - | |||||
BC052569.1 | - | |||||
BU154759.1 | - | |||||
CA406301.1 | - | |||||
CB055216.1 | - | |||||
CN314453.1 | - | |||||
CN314455.1 | - | |||||
ENST00000288840.0 | - | |||||
U59914.1 | - |